
4-(Trifluoromethyl)phenylacetic acid

Synonym(s):
(α,α,α-Trifluoro-p-tolyl)acetic acid
Linear Formula:
CF3C6H4CH2CO2H
Products
Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1