Skip to Content
MilliporeSigma
4-tert-Octylphenol-3,5-d2 monoethoxylate solution 1 μg/mL in acetone, analytical standard

4-tert-Octylphenol-3,5-d2 monoethoxylate solution

4-tert-Octylphenol-3,5-d2 monoethoxylate solution 1 μg/mL in acetone, analytical standard
Synonym(s):
2-[4-(1,1,3,3-Tetramethylbutyl)phenoxy-3,5-d2]ethanol, 4-tert-OP1EO-D2
Empirical Formula (Hill Notation):
C16D2H24O2
CAS Number:
Molecular Weight:
252.39
EC Number:

Products

Brand
Formula Weight
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique