Skip to Content
MilliporeSigma
4-tert-Butylcyclohexanecarboxylic acid 99%

4-tert-Butylcyclohexanecarboxylic acid

4-tert-Butylcyclohexanecarboxylic acid 99%
Linear Formula:
(CH3)3CC6H10CO2H
CAS Number:
Molecular Weight:
184.28

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing