Skip to Content
MilliporeSigma
4-tert-Butylbenzyl chloride 99%

4-tert-Butylbenzyl chloride

4-tert-Butylbenzyl chloride 99%
Synonym(s):
1-Chloromethyl-4-tert-butylbenzene, 4-(tert-Butyl)-α-chlorotoluene
Linear Formula:
(CH3)3CC6H4CH2Cl
CAS Number:
Molecular Weight:
182.69
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing