Skip to Content
MilliporeSigma
(4R)-2-Hydroxy-5,5-dimethyl-4-phenyl-1,3,2-dioxaphosphorinan 2-oxide 98%

(4R)-2-Hydroxy-5,5-dimethyl-4-phenyl-1,3,2-dioxaphosphorinan 2-oxide

(4R)-2-Hydroxy-5,5-dimethyl-4-phenyl-1,3,2-dioxaphosphorinan 2-oxide 98%
Synonym(s):
(2R,4R)-5,5-Dimethyl-2-hydroxy-4-phenyl-1,3,2-dioxaphosphorinan 2-oxide, (R)-(−)-2-Hydroxy-5,5-dimethyl-4-phenyl-1,3,2-dioxaphosphorinane 2-oxide
Empirical Formula (Hill Notation):
C11H15O4P
CAS Number:
Molecular Weight:
242.21

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing