Skip to Content
MilliporeSigma
4-Phenylurazole 98%

4-Phenylurazole

4-Phenylurazole 98%
Synonym(s):
4-Phenyl-1,2,4-triazolidine-3,5-dione, Ph-Ur for e-Y-CLICK
Empirical Formula (Hill Notation):
C8H7N3O2
CAS Number:
Molecular Weight:
177.16
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing