Skip to Content
MilliporeSigma
4-Nitrophenyl N-acetyl-α-D-glucosaminide ≥98%

4-Nitrophenyl N-acetyl-α-D-glucosaminide

4-Nitrophenyl N-acetyl-α-D-glucosaminide ≥98%
Synonym(s):
p-Nitrophenyl 2-acetamido-2-deoxy-α-D-glucopyranoside
Empirical Formula (Hill Notation):
C14H18N2O8
CAS Number:
Molecular Weight:
342.30
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing