Skip to Content
MilliporeSigma
4-Nitrobenzenesulfonyl fluoride 95%

4-Nitrobenzenesulfonyl fluoride

4-Nitrobenzenesulfonyl fluoride 95%
Synonym(s):
4-Nitrobenzenesulfonyl fluoride
Linear Formula:
O2NC6H4SO2F
CAS Number:
Molecular Weight:
205.16

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing