Skip to Content
MilliporeSigma
4-Methylumbelliferyl phosphate disodium salt phosphatase substrate

4-Methylumbelliferyl phosphate disodium salt

4-Methylumbelliferyl phosphate disodium salt phosphatase substrate
Synonym(s):
4-MUP, Disodium 4-methylumbelliferyl phosphate
Empirical Formula (Hill Notation):
C10H7Na2O6P
CAS Number:
Molecular Weight:
300.11
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Showing 1-1 of 1