Skip to Content
MilliporeSigma
4-Methylmorpholine N-oxide monohydrate ≥95.0% (N)

4-Methylmorpholine N-oxide monohydrate

4-Methylmorpholine N-oxide monohydrate ≥95.0% (N)
Empirical Formula (Hill Notation):
C5H11NO2 · H2O
CAS Number:
Molecular Weight:
135.16
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing