Skip to Content
MilliporeSigma
4-Methyl-1,10-phenanthroline 97%

4-Methyl-1,10-phenanthroline

4-Methyl-1,10-phenanthroline 97%
Empirical Formula (Hill Notation):
C13H10N2
CAS Number:
Molecular Weight:
194.23

Products

Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing