Skip to Content
MilliporeSigma
4-Maleimidobutyric acid N-hydroxysuccinimide ester ≥98.0% (HPLC)

4-Maleimidobutyric acid N-hydroxysuccinimide ester

4-Maleimidobutyric acid N-hydroxysuccinimide ester ≥98.0% (HPLC)
Synonym(s):
4-Maleimidobutyric acid N-succinimidyl ester, N-(γ-Maleimidobutyryloxy)succinimide, N-Succinimidyl 4-maleimidobutyrate, GMBS
Empirical Formula (Hill Notation):
C12H12N2O6
CAS Number:
Molecular Weight:
280.23

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Functional Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing