Skip to Content
MilliporeSigma
4-Hydroxy-TEMPO benzoate, free radical 97%

4-Hydroxy-TEMPO benzoate, free radical

4-Hydroxy-TEMPO benzoate, free radical 97%
Synonym(s):
4-Hydroxy-TEMPO benzoate, 4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-oxyl benzoate
Empirical Formula (Hill Notation):
C16H22NO3
CAS Number:
Molecular Weight:
276.35

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing