Skip to Content
MilliporeSigma
4-Hydroxy-TEMPO-15N 98 atom % 15N, 97% (CP)

4-Hydroxy-TEMPO-15N

4-Hydroxy-TEMPO-15N 98 atom % 15N, 97% (CP)
Synonym(s):
4-Hydroxy-2,2,6,6-tetramethylpiperidine-1-15N-oxyl, TEMPOL-15N
Empirical Formula (Hill Notation):
C9H1815NO2
CAS Number:
Molecular Weight:
173.24

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Showing 1-1 of 1
Compare
Product Number
Description
Pricing