Skip to Content
MilliporeSigma
4-Hydroxyphenylacetonitrile 98%

4-Hydroxyphenylacetonitrile

4-Hydroxyphenylacetonitrile 98%
Synonym(s):
4-Hydroxybenzyl cyanide
Linear Formula:
HOC6H4CH2CN
CAS Number:
Molecular Weight:
133.15
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing