Skip to Content
MilliporeSigma
4-(Hydroxymethyl)benzoic acid 99%

4-(Hydroxymethyl)benzoic acid

4-(Hydroxymethyl)benzoic acid 99%
Linear Formula:
HOCH2C6H4CO2H
CAS Number:
Molecular Weight:
152.15

Products

Product Category
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing