Skip to Content
MilliporeSigma
4-Hydroxymephenytoin ≥98% (HPLC)

4-Hydroxymephenytoin

4-Hydroxymephenytoin ≥98% (HPLC)
Synonym(s):
(±)-5-Ethyl-5-(4-hydroxyphenyl)-3-methylhydantoin, (±)-5-Ethyl-5-(4-hydroxyphenyl)-3-methylimididazolidine-2,4-dione
Empirical Formula (Hill Notation):
C12H14N2O3
CAS Number:
Molecular Weight:
234.25

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Research Area
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing