Skip to Content
MilliporeSigma
4-Ethynyl-1-methyl-1H-pyrazole

4-Ethynyl-1-methyl-1H-pyrazole

4-Ethynyl-1-methyl-1H-pyrazole
Empirical Formula (Hill Notation):
C6H6N2
CAS Number:
Molecular Weight:
106.13

Products

Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing