Skip to Content
MilliporeSigma
4-Ethylphenol ≥97.0% (GC)

4-Ethylphenol

4-Ethylphenol ≥97.0% (GC)
Linear Formula:
C2H5C6H4OH
CAS Number:
Molecular Weight:
122.16
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing