Skip to Content
MilliporeSigma
4-(Dimethylamino)benzylamine dihydrochloride 95%

4-(Dimethylamino)benzylamine dihydrochloride

4-(Dimethylamino)benzylamine dihydrochloride 95%
Linear Formula:
(CH3)2NC6H4CH2NH2 · 2HCl
CAS Number:
Molecular Weight:
223.14

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing