Skip to Content
MilliporeSigma
4-Chloro-7-nitrobenzofurazan BioReagent, suitable for fluorescence, ≥97.0% (HPLC)

4-Chloro-7-nitrobenzofurazan

4-Chloro-7-nitrobenzofurazan BioReagent, suitable for fluorescence, ≥97.0% (HPLC)
Synonym(s):
4-Chloro-7-nitro-1,2,3-benzoxadiazole, NBD-chloride
Empirical Formula (Hill Notation):
C6H2ClN3O3
CAS Number:
Molecular Weight:
199.55
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2