Skip to Content
MilliporeSigma
4-Chloro-2-methylthiopyrimidine 98%

4-Chloro-2-methylthiopyrimidine

4-Chloro-2-methylthiopyrimidine 98%
Empirical Formula (Hill Notation):
C5H5ClN2S
CAS Number:
Molecular Weight:
160.62
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing