Skip to Content
MilliporeSigma
4-Chloro-2-methylphenol PESTANAL®, analytical standard

4-Chloro-2-methylphenol

4-Chloro-2-methylphenol PESTANAL®, analytical standard
Synonym(s):
4-Chloro-o-cresol
Linear Formula:
ClC6H3(CH3)OH
CAS Number:
Molecular Weight:
142.58
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing