Skip to Content
MilliporeSigma
4-Bromomethylpiperidine-1-carboxylic acid tert-butyl ester 95%

4-Bromomethylpiperidine-1-carboxylic acid tert-butyl ester

4-Bromomethylpiperidine-1-carboxylic acid tert-butyl ester 95%
Synonym(s):
1-Boc-4-bromomethylpiperidine
Empirical Formula (Hill Notation):
C11H20BrNO2
CAS Number:
Molecular Weight:
278.19

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing