Skip to Content
MilliporeSigma
4-Bromomethyl-6,7-dimethoxycoumarin 96%

4-Bromomethyl-6,7-dimethoxycoumarin

4-Bromomethyl-6,7-dimethoxycoumarin 96%
Empirical Formula (Hill Notation):
C12H11BrO4
CAS Number:
Molecular Weight:
299.12

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing