Skip to Content
MilliporeSigma
4-Bromomethyl-3-nitrobenzoic acid 97%

4-Bromomethyl-3-nitrobenzoic acid

4-Bromomethyl-3-nitrobenzoic acid 97%
Linear Formula:
BrCH2C6H3(NO2)CO2H
CAS Number:
Molecular Weight:
260.04

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing