![4-Bromo-2-hydroxybenzoic acid 97%](/deepweb/assets/sigmaaldrich/product/structures/351/962/2ada5188-d066-4fc2-845c-64e8663fabf8/640/2ada5188-d066-4fc2-845c-64e8663fabf8.png)
4-Bromo-2-hydroxybenzoic acid
![4-Bromo-2-hydroxybenzoic acid 97%](/deepweb/assets/sigmaaldrich/product/structures/351/962/2ada5188-d066-4fc2-845c-64e8663fabf8/640/2ada5188-d066-4fc2-845c-64e8663fabf8.png)
Synonym(s):
4-Bromosalicylic acid
Empirical Formula (Hill Notation):
C7H5BrO3
CAS Number:
Molecular Weight:
217.02
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing