Skip to Content
MilliporeSigma
4-Bromo-1-butyne 97%

4-Bromo-1-butyne

4-Bromo-1-butyne 97%
Synonym(s):
1-Bromo-3-butyne
Empirical Formula (Hill Notation):
C4H5Br
CAS Number:
Molecular Weight:
132.99

Products

Product Category
Brand
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing