Skip to Content
MilliporeSigma
4-Benzylpiperidine 99%

4-Benzylpiperidine

4-Benzylpiperidine 99%
Synonym(s):
Phenyl(4-piperidyl)methane
Empirical Formula (Hill Notation):
C12H17N
CAS Number:
Molecular Weight:
175.27
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing