Skip to Content
MilliporeSigma
4-(Benzyloxy)aniline hydrochloride 98%

4-(Benzyloxy)aniline hydrochloride

4-(Benzyloxy)aniline hydrochloride 98%
Synonym(s):
4-Aminophenyl benzyl ether hydrochloride
Linear Formula:
C6H5CH2OC6H4NH2 · HCl
CAS Number:
Molecular Weight:
235.71
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing