Skip to Content
MilliporeSigma
4-Azidoaniline hydrochloride 97%

4-Azidoaniline hydrochloride

4-Azidoaniline hydrochloride 97%
Synonym(s):
4-Aminophenyl azide hydrochloride
Linear Formula:
N3C6H4NH2 · HCl
CAS Number:
Molecular Weight:
170.60

Products

Product Category
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing