Skip to Content
MilliporeSigma
4-Aminosalicylic acid Pharmaceutical Secondary Standard; Certified Reference Material

4-Aminosalicylic acid

4-Aminosalicylic acid Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
4-Amino-2-hydroxybenzoic acid, PAS, 4-Aminosalicylic acid
Linear Formula:
H2NC6H3-2-(OH)CO2H
CAS Number:
Molecular Weight:
153.14
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
Amino Acid
Amino Acid Type
Reaction Type
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing