Skip to Content
MilliporeSigma
4-Aminophenol PESTANAL®, analytical standard

4-Aminophenol

4-Aminophenol PESTANAL®, analytical standard
Synonym(s):
4-Hydroxyaniline, 4-Aminophenol
Linear Formula:
H2NC6H4OH
CAS Number:
Molecular Weight:
109.13
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4