Skip to Content
MilliporeSigma
4,7-Dibromobenzo[c]-1,2,5-thiadiazole 95%

4,7-Dibromobenzo[c]-1,2,5-thiadiazole

4,7-Dibromobenzo[c]-1,2,5-thiadiazole 95%
Empirical Formula (Hill Notation):
C6H2Br2N2S
CAS Number:
Molecular Weight:
293.97

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing