Skip to Content
MilliporeSigma
4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosane 98%

4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosane

4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosane 98%
Synonym(s):
Kryptofix® 221
Empirical Formula (Hill Notation):
C16H32N2O5
CAS Number:
Molecular Weight:
332.44
EC Number:

Products

Brand
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing