Skip to Content
MilliporeSigma
4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane 98%

4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane

4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane 98%
Empirical Formula (Hill Notation):
C18H36N2O6
CAS Number:
Molecular Weight:
376.49
EC Number:

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing