Skip to Content
MilliporeSigma
4′,6-Diamidino-2-phenylindole dihydrochloride suitable for fluorescence, BioReagent, ≥95.0% (HPLC)

4′,6-Diamidino-2-phenylindole dihydrochloride

4′,6-Diamidino-2-phenylindole dihydrochloride suitable for fluorescence, BioReagent, ≥95.0% (HPLC)
Synonym(s):
2-(4-Amidinophenyl)-6-indolecarbamidine dihydrochloride, DAPI dihydrochloride
Empirical Formula (Hill Notation):
C16H15N5 · 2HCl
CAS Number:
Molecular Weight:
350.25
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Technique