Skip to Content
MilliporeSigma
4-(4-Hydroxyphenyl)-2-butanone ≥98%, FCC, FG

4-(4-Hydroxyphenyl)-2-butanone

4-(4-Hydroxyphenyl)-2-butanone ≥98%, FCC, FG
Synonym(s):
Frambione, NSC 26515, Raspberry ketone
Linear Formula:
HOC6H4CH2CH2COCH3
CAS Number:
Molecular Weight:
164.20
EC Number:

Products

Product Category
Sustainability
Biological Source
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-3 of 3