Skip to Content
MilliporeSigma
4-(4-Hydroxy-4-methylpentyl)-3-cyclohexene-1-carboxaldehyde mixture of isomers, ≥97.0% (GC)

4-(4-Hydroxy-4-methylpentyl)-3-cyclohexene-1-carboxaldehyde

4-(4-Hydroxy-4-methylpentyl)-3-cyclohexene-1-carboxaldehyde mixture of isomers, ≥97.0% (GC)
Empirical Formula (Hill Notation):
C13H22O2
CAS Number:
Molecular Weight:
210.31
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1