Skip to Content
MilliporeSigma
4-(4-Chlorophenyl)-4-hydroxypiperidine certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

4-(4-Chlorophenyl)-4-hydroxypiperidine

4-(4-Chlorophenyl)-4-hydroxypiperidine certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Haloperidol metabolite I, 4-(4-Chlorophenyl)-4-hydroxypiperidine, 4-(4-Chlorophenyl)-4-piperidinol
Empirical Formula (Hill Notation):
C11H14ClNO
CAS Number:
Molecular Weight:
211.69

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade