Skip to Content
MilliporeSigma
4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-Heptadecafluoroundecanoic acid ≥95.0% (NT)

4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-Heptadecafluoroundecanoic acid

4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-Heptadecafluoroundecanoic acid ≥95.0% (NT)
Synonym(s):
2H,2H,3H,3H-Perfluoroundecanoic acid
Linear Formula:
CF3(CF2)7CH2CH2COOH
CAS Number:
Molecular Weight:
492.13
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1