Skip to Content
MilliporeSigma
4,4,4-Trifluoro-1-(2-naphthyl)-1,3-butanedione 99%

4,4,4-Trifluoro-1-(2-naphthyl)-1,3-butanedione

4,4,4-Trifluoro-1-(2-naphthyl)-1,3-butanedione 99%
Synonym(s):
4,4,4-Trifluoro-3-oxo-2′-butyronaphthone, 1-(2-Naphthoyl)-3,3,3-trifluoroacetone, 2-(4,4,4-Trifluoroacetoacetyl)naphthalene
Linear Formula:
C10H7COCH2COCF3
CAS Number:
Molecular Weight:
266.22

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing