Skip to Content
MilliporeSigma
4-[4-(4-Chlorophenyl)-3,6-dihydropyridin-1(2H)-yl]-N,N-dimethyl-2,2-diphenylbutanamide certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

4-[4-(4-Chlorophenyl)-3,6-dihydropyridin-1(2H)-yl]-N,N-dimethyl-2,2-diphenylbutanamide

4-[4-(4-Chlorophenyl)-3,6-dihydropyridin-1(2H)-yl]-N,N-dimethyl-2,2-diphenylbutanamide certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
4-(4-Chlorophenyl)-3,6-dihydro-N,N-dimethyl-α,α-diphenyl-1(2H)-pyridinebutanamide
Empirical Formula (Hill Notation):
C29H31ClN2O
CAS Number:
Molecular Weight:
459.02

Products

Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade