Skip to Content
MilliporeSigma
4,4′-(1-Phenylethylidene)bisphenol 99%

4,4′-(1-Phenylethylidene)bisphenol

4,4′-(1-Phenylethylidene)bisphenol 99%
Synonym(s):
Bisphenol AP
Linear Formula:
CH3C(C6H5)(C6H4OH)2
CAS Number:
Molecular Weight:
290.36

Products

Product Category
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing