Skip to Content
MilliporeSigma
4-(3-Chloropropyl)morpholine hydrochloride 97%

4-(3-Chloropropyl)morpholine hydrochloride

4-(3-Chloropropyl)morpholine hydrochloride 97%
Empirical Formula (Hill Notation):
C7H14ClNO · HCl
CAS Number:
Molecular Weight:
200.11
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing