Skip to Content
MilliporeSigma
4-(2-Aminoethyl)aniline 97%

4-(2-Aminoethyl)aniline

4-(2-Aminoethyl)aniline 97%
Synonym(s):
4-Aminophenethylamine
Linear Formula:
H2NC6H4CH2CH2NH2
CAS Number:
Molecular Weight:
136.19
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing