Skip to Content
MilliporeSigma
4-(1′,5′-Dihydro-1′-methyl-2′H-[5,6]fullereno-C60-Ih-[1,9-c]pyrrol-2′-yl)benzoic acid

4-(1′,5′-Dihydro-1′-methyl-2′H-[5,6]fullereno-C60-Ih-[1,9-c]pyrrol-2′-yl)benzoic acid

4-(1′,5′-Dihydro-1′-methyl-2′H-[5,6]fullereno-C60-Ih-[1,9-c]pyrrol-2′-yl)benzoic acid
Empirical Formula (Hill Notation):
C70H11NO2
CAS Number:
Molecular Weight:
897.84

Products

Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1