Skip to Content
MilliporeSigma
3-Ureidopropionic acid ≥98.0% (T)

3-Ureidopropionic acid

3-Ureidopropionic acid ≥98.0% (T)
Synonym(s):
N-Carbamoyl-β-alanine
Linear Formula:
NH2CONHCH2CH2COOH
CAS Number:
Molecular Weight:
132.12

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing