Skip to Content
MilliporeSigma
3-(Triethoxysilyl)propyl isocyanate 95%

3-(Triethoxysilyl)propyl isocyanate

3-(Triethoxysilyl)propyl isocyanate 95%
Synonym(s):
Triethoxy(3-isocyanatopropyl)silane, (3-Isocyanatopropyl)triethoxysilane
Linear Formula:
(C2H5O)3Si(CH2)3NCO
CAS Number:
Molecular Weight:
247.36
EC Number:

Products

Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing