Skip to Content
MilliporeSigma
3-Nitrophthalic anhydride 98%

3-Nitrophthalic anhydride

3-Nitrophthalic anhydride 98%
Synonym(s):
3-Nitrophthalic anhydride
Empirical Formula (Hill Notation):
C8H3NO5
CAS Number:
Molecular Weight:
193.11

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing